Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGATATACACACCTAGCCTCAGTC[C/T]CCCACCCCCAGTCCTGCCATTGGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609921 MIM: 616955 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LRP10 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LRP10 - LDL receptor related protein 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005267510.1 | 1358 | Missense Mutation | CCC,TCC | P,S 223 | XP_005267567.1 |
REM2 - RRAD and GEM like GTPase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |