Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTGCCTCGATGTTCCAGCTGCCCA[C/T]CTTGAATTTCAGCCCCCAGCAAGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606326 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C14orf39 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C14orf39 - chromosome 14 open reading frame 39 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SIX6 - SIX homeobox 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007374.2 | 196 | Missense Mutation | ACC,ATC | T,I 6 | NP_031400.2 |