Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTGCATAGATGCCTCCATATCGC[C/T]GTGCATGCTTCCATGTGCCAAACTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 107748 MIM: 610107 MIM: 609865 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
APEX1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
APEX1 - apurinic/apyrimidinic endodeoxyribonuclease 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001244249.1 | 1021 | Intron | NP_001231178.1 | |||
NM_001641.3 | 1021 | Intron | NP_001632.2 | |||
NM_080648.2 | 1021 | Intron | NP_542379.1 | |||
NM_080649.2 | 1021 | Intron | NP_542380.1 |
OSGEP - O-sialoglycoprotein endopeptidase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM55B - transmembrane protein 55B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001100814.2 | 1021 | Missense Mutation | CAG,CGG | Q,R 245 | NP_001094284.1 | |
NM_144568.3 | 1021 | Missense Mutation | CAG,CGG | Q,R 238 | NP_653169.2 |