Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGTCCTGCTTTGGCTGCCAGGGC[G/T]TCATTCTGCTGAATATGATATTCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607384 | ||||||||||||||||||||
Literature Links: |
TIMM9 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TIMM9 - translocase of inner mitochondrial membrane 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304485.1 | 356 | Missense Mutation | GAA,GAC | E,D 76 | NP_001291414.1 | |
NM_001304486.1 | 356 | Missense Mutation | GAA,GAC | E,D 76 | NP_001291415.1 | |
NM_001304487.1 | 356 | Missense Mutation | GAA,GAC | E,D 76 | NP_001291416.1 | |
NM_001304488.1 | 356 | Missense Mutation | GAA,GAC | E,D 53 | NP_001291417.1 | |
NM_001304489.1 | 356 | Missense Mutation | GAA,GAC | E,D 48 | NP_001291418.1 | |
NM_001304490.1 | 356 | Missense Mutation | GAA,GAC | E,D 47 | NP_001291419.1 | |
NM_001304491.1 | 356 | Missense Mutation | GAA,GAC | E,D 47 | NP_001291420.1 | |
NM_012460.3 | 356 | Missense Mutation | GAA,GAC | E,D 76 | NP_036592.1 |
TOMM20L - translocase of outer mitochondrial membrane 20 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_207377.2 | 356 | Intron | NP_997260.1 | |||
XM_011536742.2 | 356 | Intron | XP_011535044.1 | |||
XM_011536743.2 | 356 | Intron | XP_011535045.1 | |||
XM_011536744.2 | 356 | Intron | XP_011535046.1 |