Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGTGAAGAAAAACCTGGCGTTGAA[C/T]GTGTACATAAATTGTGGTAATGAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C14orf79 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C14orf79 - chromosome 14 open reading frame 79 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_174891.3 | 706 | Missense Mutation | CGT,TGT | R,C 185 | NP_777551.2 | |
XM_005267317.3 | 706 | Missense Mutation | CGT,TGT | R,C 185 | XP_005267374.1 | |
XM_005267318.3 | 706 | Missense Mutation | CGT,TGT | R,C 185 | XP_005267375.1 |