Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAAGAAAGAAAAGAAGGATCCCAG[C/T]GCATTAAAGGAAAGAGAAGTGTGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615380 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AREL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AREL1 - apoptosis resistant E3 ubiquitin protein ligase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FCF1 - FCF1 rRNA-processing protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318508.1 | 176 | Intron | NP_001305437.1 | |||
NM_015962.4 | 176 | Intron | NP_057046.1 | |||
XM_005267730.3 | 176 | UTR 5 | XP_005267787.1 | |||
XM_011536815.2 | 176 | Intron | XP_011535117.1 | |||
XM_011536816.2 | 176 | Intron | XP_011535118.1 |