Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCTCGTTCTCAGGGATGCTGCTGA[A/G]GTTAAAGAGGAAACGAAAAGCAGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 112262 | ||||||||||||||||||||
Literature Links: |
BMP4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BMP4 - bone morphogenetic protein 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001202.4 | 655 | Missense Mutation | CTC,TTC | L,F 144 | NP_001193.2 | |
NM_130850.3 | 655 | Missense Mutation | CTC,TTC | L,F 144 | NP_570911.2 | |
XM_005268015.4 | 655 | Missense Mutation | CTC,TTC | L,F 144 | XP_005268072.1 | |
XM_017021603.1 | 655 | Missense Mutation | CTC,TTC | L,F 144 | XP_016877092.1 | |
XM_017021604.1 | 655 | Missense Mutation | CTC,TTC | L,F 81 | XP_016877093.1 | |
XM_017021605.1 | 655 | Missense Mutation | CTC,TTC | L,F 81 | XP_016877094.1 |
MIR5580 - microRNA 5580 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |