Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCATCTCTTCCTTTGCTGTCTGAG[C/T]TCCTTTGGAATGCATCCCCACCTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609798 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NEK9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NEK9 - NIMA related kinase 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005268208.1 | 2813 | Missense Mutation | ACT,GCT | T,A 959 | XP_005268265.1 | |
XM_005268209.3 | 2813 | Missense Mutation | ACT,GCT | T,A 841 | XP_005268266.1 | |
XM_017021765.1 | 2813 | Missense Mutation | ACT,GCT | T,A 829 | XP_016877254.1 |
ZC2HC1C - zinc finger C2HC-type containing 1C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |