Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTAGATGGCCGCCCTGCTCCCCCA[A/G]TGAAAGGCCAGCTTCGAAGAAAAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600754 MIM: 611856 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MMP14 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MMP14 - matrix metallopeptidase 14 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MRPL52 - mitochondrial ribosomal protein L52 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318870.1 | 291 | Missense Mutation | ATG,GTG | M,V 59 | NP_001305799.1 | |
NM_178336.2 | 291 | Missense Mutation | ATG,GTG | M,V 60 | NP_848026.1 | |
NM_180982.2 | 291 | Missense Mutation | ATG,GTG | M,V 59 | NP_851313.1 | |
NM_181304.2 | 291 | Missense Mutation | ATG,GTG | M,V 1 | NP_851821.1 | |
NM_181305.2 | 291 | Missense Mutation | ATG,GTG | M,V 1 | NP_851822.1 | |
NM_181306.2 | 291 | Missense Mutation | ATG,GTG | M,V 1 | NP_851823.1 | |
NM_181307.2 | 291 | Intron | NP_851824.1 | |||
XM_005267325.4 | 291 | Missense Mutation | ATG,GTG | M,V 60 | XP_005267382.1 | |
XM_005267328.3 | 291 | Missense Mutation | ATG,GTG | M,V 1 | XP_005267385.1 |