Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACGCTGTCCAAGCCTGACAGCCCG[C/T]GGCCTGACTTTGCGGTGGAGACGTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615364 | ||||||||||||||||||||
Literature Links: |
AK7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AK7 - adenylate kinase 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152327.3 | 309 | Missense Mutation | CGG,TGG | R,W 89 | NP_689540.2 | |
XM_006720021.2 | 309 | Missense Mutation | CGG,TGG | R,W 89 | XP_006720084.1 | |
XM_006720022.2 | 309 | Missense Mutation | CGG,TGG | R,W 89 | XP_006720085.1 | |
XM_011536403.2 | 309 | Missense Mutation | CGG,TGG | R,W 89 | XP_011534705.1 | |
XM_017020956.1 | 309 | Missense Mutation | CGG,TGG | R,W 89 | XP_016876445.1 | |
XM_017020957.1 | 309 | Missense Mutation | CGG,TGG | R,W 89 | XP_016876446.1 | |
XM_017020958.1 | 309 | Missense Mutation | CGG,TGG | R,W 89 | XP_016876447.1 | |
XM_017020959.1 | 309 | Missense Mutation | CGG,TGG | R,W 89 | XP_016876448.1 | |
XM_017020960.1 | 309 | Missense Mutation | CGG,TGG | R,W 89 | XP_016876449.1 |