Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTCAGCTGTTTTGAGAAGCCTTC[A/G]GGTGAATGTGGGTCCAGACGGAGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF410 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF410 - zinc finger protein 410 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242924.1 | 686 | Missense Mutation | CAG,CGG | Q,R 96 | NP_001229853.1 | |
NM_001242926.1 | 686 | Missense Mutation | CAG,CGG | Q,R 79 | NP_001229855.1 | |
NM_001242927.1 | 686 | Intron | NP_001229856.1 | |||
NM_001242928.1 | 686 | UTR 5 | NP_001229857.1 | |||
NM_021188.2 | 686 | Missense Mutation | CAG,CGG | Q,R 79 | NP_067011.1 |