Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTTAAGCCCGGGAGAGAATAGTT[G/T]GAGCTCCTGGCGTGGCCCATATTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601205 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105378189 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC105378189 - uncharacterized LOC105378189 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SALRNA1 - senescence associated long non-coding RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SIX1 - SIX homeobox 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005982.3 | 973 | Silent Mutation | TCA,TCC | S,S 244 | NP_005973.1 | |
XM_017021602.1 | 973 | UTR 3 | XP_016877091.1 |