Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAACTGACATGAAAGACATGAGGCT[C/T]GAAGCTGAAGCAGTTGTAAATGATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615364 MIM: 616605 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AK7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AK7 - adenylate kinase 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GSKIP - GSK3B interacting protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271904.1 | 223 | Silent Mutation | CTC,CTT | L,L 35 | NP_001258833.1 | |
NM_001271905.1 | 223 | Silent Mutation | CTC,CTT | L,L 35 | NP_001258834.1 | |
NM_001271906.1 | 223 | Silent Mutation | CTC,CTT | L,L 35 | NP_001258835.1 | |
NM_016472.4 | 223 | Silent Mutation | CTC,CTT | L,L 35 | NP_057556.2 | |
XM_017021365.1 | 223 | Silent Mutation | CTC,CTT | L,L 35 | XP_016876854.1 |