Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCACACATGTTCTTTCCGCGCTCGA[G/T]GAGGAAGTACCTGGGCAGAAAGAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 116820 | ||||||||||||||||||||
Literature Links: |
CTSH PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CTSH - cathepsin H | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319137.1 | 1060 | Missense Mutation | NP_001306066.1 | |||
NM_004390.4 | 1060 | Missense Mutation | NP_004381.2 | |||
XM_005254181.2 | 1060 | Missense Mutation | XP_005254238.1 | |||
XM_011521276.1 | 1060 | Intron | XP_011519578.1 | |||
XM_017021951.1 | 1060 | Missense Mutation | XP_016877440.1 | |||
XM_017021952.1 | 1060 | Missense Mutation | XP_016877441.1 |