Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATACTGGTATGTGCAAAATGGACA[C/T]TCTCATCCAGGAAACCATGTAAGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606819 | ||||||||||||||||||||
Literature Links: |
DPP8 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DPP8 - dipeptidyl peptidase 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320875.1 | 1377 | Missense Mutation | AAT,AGT | N,S 835 | NP_001307804.1 | |
NM_001320876.1 | 1377 | Missense Mutation | AAT,AGT | N,S 819 | NP_001307805.1 | |
NM_017743.5 | 1377 | Missense Mutation | AAT,AGT | N,S 719 | NP_060213.2 | |
NM_130434.4 | 1377 | Missense Mutation | AAT,AGT | N,S 819 | NP_569118.1 | |
NM_197960.3 | 1377 | Missense Mutation | AAT,AGT | N,S 835 | NP_932064.1 | |
NM_197961.3 | 1377 | Missense Mutation | AAT,AGT | N,S 784 | NP_932065.1 | |
XM_005254505.1 | 1377 | Missense Mutation | AAT,AGT | N,S 416 | XP_005254562.1 | |
XM_011521730.1 | 1377 | Missense Mutation | AAT,AGT | N,S 792 | XP_011520032.1 | |
XM_011521731.1 | 1377 | Missense Mutation | AAT,AGT | N,S 777 | XP_011520033.1 | |
XM_011521732.2 | 1377 | Intron | XP_011520034.1 | |||
XM_011521733.1 | 1377 | Missense Mutation | AAT,AGT | N,S 761 | XP_011520035.1 | |
XM_011521734.1 | 1377 | Missense Mutation | AAT,AGT | N,S 735 | XP_011520036.1 | |
XM_017022373.1 | 1377 | Missense Mutation | AAT,AGT | N,S 835 | XP_016877862.1 | |
XM_017022374.1 | 1377 | Missense Mutation | AAT,AGT | N,S 784 | XP_016877863.1 | |
XM_017022375.1 | 1377 | Missense Mutation | AAT,AGT | N,S 776 | XP_016877864.1 | |
XM_017022376.1 | 1377 | Missense Mutation | AAT,AGT | N,S 768 | XP_016877865.1 | |
XM_017022377.1 | 1377 | Missense Mutation | AAT,AGT | N,S 718 | XP_016877866.1 | |
XM_017022378.1 | 1377 | Intron | XP_016877867.1 | |||
XM_017022379.1 | 1377 | Missense Mutation | AAT,AGT | N,S 710 | XP_016877868.1 | |
XM_017022380.1 | 1377 | Intron | XP_016877869.1 | |||
XM_017022381.1 | 1377 | Missense Mutation | AAT,AGT | N,S 667 | XP_016877870.1 | |
XM_017022382.1 | 1377 | Missense Mutation | AAT,AGT | N,S 451 | XP_016877871.1 | |
XM_017022383.1 | 1377 | Missense Mutation | AAT,AGT | N,S 451 | XP_016877872.1 |