Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATGCCAGCCAGGGCCAGTGAGAGG[A/T]TGATGCCTGGTGAAGAGGTAGAGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603263 | ||||||||||||||||||||
Literature Links: |
CA12 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CA12 - carbonic anhydrase 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001218.4 | 1271 | Missense Mutation | AAC,ATC | N,I 305 | NP_001209.1 | |
NM_001293642.1 | 1271 | Missense Mutation | AAC,ATC | N,I 234 | NP_001280571.1 | |
NM_206925.2 | 1271 | Missense Mutation | AAC,ATC | N,I 294 | NP_996808.1 |