Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATCTTCTTGTAATATTGAACTGAA[A/G]CCTTTGGATAGCGAGGCTTATTTCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609984 | ||||||||||||||||||||
Literature Links: |
LCTL PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LCTL - lactase like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278562.1 | 1598 | Missense Mutation | GCT,GTT | A,V 316 | NP_001265491.1 | |
NM_207338.3 | 1598 | Missense Mutation | GCT,GTT | A,V 489 | NP_997221.2 | |
XM_017021999.1 | 1598 | Missense Mutation | GCT,GTT | A,V 506 | XP_016877488.1 |
ZWILCH - zwilch kinetochore protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |