Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGTTGCCCAAGGGGGGAAGCAGCC[A/G]CTTCTTGCTGTTTCGGGTCTCTGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612183 | ||||||||||||||||||||
Literature Links: |
GPR176 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GPR176 - G protein-coupled receptor 176 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271854.1 | 2609 | Silent Mutation | NP_001258783.1 | |||
NM_001271855.1 | 2609 | Silent Mutation | NP_001258784.1 | |||
NM_007223.2 | 2609 | Silent Mutation | NP_009154.1 | |||
XM_011521167.2 | 2609 | Missense Mutation | XP_011519469.1 | |||
XM_011521168.2 | 2609 | Missense Mutation | XP_011519470.1 | |||
XM_017021873.1 | 2609 | Missense Mutation | XP_016877362.1 | |||
XM_017021874.1 | 2609 | Missense Mutation | XP_016877363.1 | |||
XM_017021875.1 | 2609 | Silent Mutation | XP_016877364.1 | |||
XM_017021876.1 | 2609 | Missense Mutation | XP_016877365.1 | |||
XM_017021877.1 | 2609 | Missense Mutation | XP_016877366.1 | |||
XM_017021878.1 | 2609 | Missense Mutation | XP_016877367.1 |