Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGAGCCACTGGCTACATCTTGTCCT[A/G]CAGGTAAGAGGGGAAGCCCTCTGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
SH2D7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SH2D7 - SH2 domain containing 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001101404.1 | 263 | Missense Mutation | TAC,TGC | Y,C 88 | NP_001094874.1 | |
XM_011521895.2 | 263 | Missense Mutation | TAC,TGC | Y,C 88 | XP_011520197.1 | |
XM_011521896.2 | 263 | Missense Mutation | ACA,GCA | T,A 36 | XP_011520198.1 | |
XM_011521899.2 | 263 | Intron | XP_011520201.1 | |||
XM_017022487.1 | 263 | Missense Mutation | ACA,GCA | T,A 50 | XP_016877976.1 | |
XM_017022488.1 | 263 | UTR 5 | XP_016877977.1 |