Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAACCAAGAAGGATGTACAATGAT[A/T]AGGGATTTTAGATTTTTCCGTAACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603292 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BNIP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BNIP2 - BCL2 interacting protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320674.1 | 1189 | Silent Mutation | CTA,CTT | L,L 365 | NP_001307603.1 | |
NM_001320675.1 | 1189 | Silent Mutation | CTA,CTT | L,L 306 | NP_001307604.1 | |
NM_004330.3 | 1189 | Silent Mutation | CTA,CTT | L,L 365 | NP_004321.2 |