Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCCTGGGCAACCTCAGCACCCCTA[A/C]GGGTAGCCCATCTGCAGTCTGCATC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611300 | ||||||||||||||||||||
Literature Links: |
LARP6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LARP6 - La ribonucleoprotein domain family member 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286679.1 | 1308 | Missense Mutation | NP_001273608.1 | |||
NM_018357.3 | 1308 | Missense Mutation | NP_060827.2 | |||
NM_197958.2 | 1308 | Intron | NP_932062.1 |