Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCTTCCGGGACTCCAGATCAACGA[A/C]CTGCAGACTTCGGAGCCTCGCCCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 162340 MIM: 610417 MIM: 616144 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NMB PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NMB - neuromedin B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SCAND2P - SCAN domain containing 2 pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
WDR73 - WD repeat domain 73 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032856.3 | 547 | Missense Mutation | GTC,TTC | V,F 160 | NP_116245.2 |