Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGGGTGGGCCACAGCTTGCTGCA[C/G]GTCTTCAGTACCGAGTTCCGAGCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C15orf57 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C15orf57 - chromosome 15 open reading frame 57 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RPUSD2 - RNA pseudouridylate synthase domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286407.1 | 537 | Intron | NP_001273336.1 | |||
NM_152260.2 | 537 | Silent Mutation | CAC,CAG | H,Q 164 | NP_689473.1 |