Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCAGTGGGGACTTTCTGGAAACGC[C/T]TTTGCGTTGGGCCAAGGGGCTGCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611300 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LARP6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LARP6 - La ribonucleoprotein domain family member 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286679.1 | 1083 | Missense Mutation | AGC,GGC | S,G 207 | NP_001273608.1 | |
NM_018357.3 | 1083 | Missense Mutation | AGC,GGC | S,G 391 | NP_060827.2 | |
NM_197958.2 | 1083 | Intron | NP_932062.1 |