Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCCAACCGGCTGCCAGGCTTGCC[G/T]GAGGGTGCCTTCAGGGAGGTGCCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602059 MIM: 610745 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ISLR PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ISLR - immunoglobulin superfamily containing leucine rich repeat | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005545.3 | 480 | Silent Mutation | CCG,CCT | P,P 66 | NP_005536.1 | |
NM_201526.1 | 480 | Silent Mutation | CCG,CCT | P,P 66 | NP_958934.1 |
STRA6 - stimulated by retinoic acid 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |