Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAAAGGGTTTCTCGTCATAATGGC[C/T]GTGGAAGGGTCAACCATTACCAGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616786 MIM: 616061 | ||||||||||||||||||||
Literature Links: |
MAPKBP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MAPKBP1 - mitogen-activated protein kinase binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128608.1 | 256 | Silent Mutation | GCC,GCT | A,A 2 | NP_001122080.1 | |
NM_001265611.1 | 256 | Silent Mutation | GCC,GCT | A,A 2 | NP_001252540.1 | |
NM_014994.2 | 256 | Silent Mutation | GCC,GCT | A,A 2 | NP_055809.2 | |
XM_006720438.1 | 256 | Silent Mutation | GCC,GCT | A,A 2 | XP_006720501.1 | |
XM_006720439.2 | 256 | Intron | XP_006720502.1 | |||
XM_011521383.1 | 256 | Silent Mutation | GCC,GCT | A,A 2 | XP_011519685.1 | |
XM_011521384.2 | 256 | Silent Mutation | GCC,GCT | A,A 2 | XP_011519686.1 | |
XM_017022017.1 | 256 | Silent Mutation | GCC,GCT | A,A 2 | XP_016877506.1 |
MGA - MGA, MAX dimerization protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |