Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGACAAAAGGGGGCCAAACATGGGC[A/G]AGAAGGGCTCTGTTGCTCGGCATCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609802 | ||||||||||||||||||||
Literature Links: |
SLC24A5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SLC24A5 - solute carrier family 24 member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_205850.2 | 103 | Silent Mutation | GCA,GCG | A,A 10 | NP_995322.1 | |
XM_017022079.1 | 103 | Intron | XP_016877568.1 | |||
XM_017022080.1 | 103 | Intron | XP_016877569.1 |