Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTCCTGACCAACCACTGAAGGCA[C/T]TGGCCTATGTGGCCACCCCACAGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614587 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHAC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CHAC1 - ChaC glutathione specific gamma-glutamylcyclotransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142776.1 | 567 | Intron | NP_001136248.1 | |||
NM_024111.3 | 567 | Silent Mutation | CTG,TTG | L,L 183 | NP_077016.2 | |
XM_017022563.1 | 567 | Silent Mutation | CTG,TTG | L,L 183 | XP_016878052.1 | |
XM_017022564.1 | 567 | Silent Mutation | CTG,TTG | L,L 183 | XP_016878053.1 | |
XM_017022565.1 | 567 | Silent Mutation | CTG,TTG | L,L 183 | XP_016878054.1 |