Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGTGGGAGAACAACGGGCAGGTGT[A/G]CAGCTTGCTCAACTCGGGCTCAGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 153456 MIM: 616800 | ||||||||||||||||||||
Literature Links: |
LOXL1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOXL1 - lysyl oxidase like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005576.3 | 488 | Missense Mutation | TAC,TGC | Y,C 52 | NP_005567.2 | |
XM_011521555.1 | 488 | Missense Mutation | TAC,TGC | Y,C 52 | XP_011519857.1 | |
XM_017022179.1 | 488 | Intron | XP_016877668.1 |
LOXL1-AS1 - LOXL1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |