Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCAGAGCAAATGGCATTCGTTGG[C/T]TTCAAAGGCAGCTTCCGGCCCATCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608366 MIM: 607783 | ||||||||||||||||||||
Literature Links: |
CEMIP PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CEMIP - cell migration inducing hyaluronan binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001293298.1 | 4266 | Silent Mutation | GGC,GGT | G,G 1326 | NP_001280227.1 | |
NM_001293304.1 | 4266 | Silent Mutation | GGC,GGT | G,G 1326 | NP_001280233.1 | |
NM_018689.2 | 4266 | Silent Mutation | GGC,GGT | G,G 1326 | NP_061159.1 |
MESDC2 - mesoderm development candidate 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015154.2 | 4266 | Intron | NP_055969.1 |