Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTATCGAAGGTGTATTCCACCTCA[C/T]AGAAGCTGACAATGGTCATGGCGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616438 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PIRC66 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PIRC66 - piwi-interacting RNA cluster 66 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TNFAIP8L3 - TNF alpha induced protein 8 like 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001311175.1 | 831 | Missense Mutation | TAT,TGT | Y,C 123 | NP_001298104.1 | |
NM_207381.3 | 831 | Missense Mutation | TAT,TGT | Y,C 211 | NP_997264.2 | |
XM_011521545.2 | 831 | Missense Mutation | TAT,TGT | Y,C 137 | XP_011519847.1 | |
XM_017022168.1 | 831 | Missense Mutation | TAT,TGT | Y,C 122 | XP_016877657.1 | |
XM_017022169.1 | 831 | Missense Mutation | TAT,TGT | Y,C 77 | XP_016877658.1 |