Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCATTTTTCAGACTCCAAACCCC[A/T]TGATGAGCTCTTGCAGAACAACAGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605907 | ||||||||||||||||||||
Literature Links: |
ALG1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ALG1 - ALG1, chitobiosyldiphosphodolichol beta-mannosyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019109.4 | 262 | Missense Mutation | CAT,CTT | H,L 74 | NP_061982.3 | |
XM_011522565.1 | 262 | UTR 5 | XP_011520867.1 | |||
XM_017023457.1 | 262 | Missense Mutation | CAT,CTT | H,L 74 | XP_016878946.1 | |
XM_017023458.1 | 262 | UTR 5 | XP_016878947.1 |
C16orf89 - chromosome 16 open reading frame 89 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |