Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600227 MIM: 602349 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C16orf59 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C16orf59 - chromosome 16 open reading frame 59 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_025108.2 | 669 | Missense Mutation | CAG,CGG | Q,R 245 | NP_079384.2 | |
XM_011522667.1 | 669 | Missense Mutation | CAG,CGG | Q,R 212 | XP_011520969.1 |
CCNF - cyclin F | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC729652 - uncharacterized LOC729652 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR6768 - microRNA 6768 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NTN3 - netrin 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |