Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCTCTCCTCTTTCTGAGGACAGC[C/T]TCTCCTTCAGGAGTTTCCCGGTCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM65A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM65A - family with sequence similarity 65 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193522.1 | 227 | Missense Mutation | CTC,TTC | L,F 36 | NP_001180451.1 | |
NM_001193523.1 | 227 | Intron | NP_001180452.1 | |||
NM_001193524.1 | 227 | Intron | NP_001180453.1 | |||
NM_024519.3 | 227 | Intron | NP_078795.2 | |||
XM_011523321.1 | 227 | Intron | XP_011521623.1 | |||
XM_011523322.1 | 227 | Intron | XP_011521624.1 | |||
XM_011523324.2 | 227 | Intron | XP_011521626.1 | |||
XM_011523325.1 | 227 | Intron | XP_011521627.1 | |||
XM_011523326.1 | 227 | Intron | XP_011521628.1 | |||
XM_017023663.1 | 227 | Intron | XP_016879152.1 | |||
XM_017023664.1 | 227 | Intron | XP_016879153.1 |
LOC100505942 - uncharacterized LOC100505942 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |