Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCACTCACCCATGTACTGCTTCAT[C/G]TGCTGCCTGGCCTTGTCGGACCTGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 155555 MIM: 612326 MIM: 602661 | ||||||||||||||||||||
Literature Links: |
MC1R PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MC1R - melanocortin 1 receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002386.3 | 1611 | Missense Mutation | ATC,ATG | I,M 77 | NP_002377.4 |
TCF25 - transcription factor 25 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TUBB3 - tubulin beta 3 class III | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |