Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGGCCTACTACACCCGTTACTTC[A/G]TATCGAATATCTATCTGAGCCCTCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608159 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRSS21 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PRSS21 - protease, serine 21 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270452.1 | 387 | Missense Mutation | ATA,GTA | I,V 119 | NP_001257381.1 | |
NM_006799.3 | 387 | Missense Mutation | ATA,GTA | I,V 121 | NP_006790.1 | |
NM_144956.2 | 387 | Missense Mutation | ATA,GTA | I,V 119 | NP_659205.1 | |
NM_144957.2 | 387 | Missense Mutation | ATA,GTA | I,V 121 | NP_659206.1 | |
XM_017022875.1 | 387 | Missense Mutation | ATA,GTA | I,V 119 | XP_016878364.1 | |
XM_017022876.1 | 387 | Missense Mutation | ATA,GTA | I,V 121 | XP_016878365.1 | |
XM_017022877.1 | 387 | Missense Mutation | ATA,GTA | I,V 119 | XP_016878366.1 | |
XM_017022878.1 | 387 | Missense Mutation | ATA,GTA | I,V 121 | XP_016878367.1 |