Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGCCCTGCTACCTGCGCATGGAC[A/G]CACTGGCGCTGCTTGGGGGACTGGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609899 MIM: 614578 MIM: 602474 | ||||||||||||||||||||
Literature Links: |
KREMEN2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KREMEN2 - kringle containing transmembrane protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PAQR4 - progestin and adipoQ receptor family member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001284511.1 | 944 | Missense Mutation | ACA,GCA | T,A 172 | NP_001271440.1 | |
NM_001284512.1 | 944 | Missense Mutation | ACA,GCA | T,A 137 | NP_001271441.1 | |
NM_001284513.1 | 944 | Missense Mutation | ACA,GCA | T,A 144 | NP_001271442.1 | |
NM_001324118.1 | 944 | Intron | NP_001311047.1 | |||
NM_152341.4 | 944 | Missense Mutation | ACA,GCA | T,A 211 | NP_689554.2 |
PKMYT1 - protein kinase, membrane associated tyrosine/threonine 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |