Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCCATGGTACAGCTGGCTCCTGC[G/T]GCAGCCATGGACGAGGTCACCTTTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615261 | ||||||||||||||||||||
Literature Links: |
METTL22 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
METTL22 - methyltransferase like 22 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024109.3 | 282 | Silent Mutation | GCG,GCT | A,A 7 | NP_077014.3 | |
XM_005255570.3 | 282 | Silent Mutation | GCG,GCT | A,A 7 | XP_005255627.1 | |
XM_005255571.2 | 282 | Silent Mutation | GCG,GCT | A,A 7 | XP_005255628.1 | |
XM_011522657.2 | 282 | Silent Mutation | GCG,GCT | A,A 7 | XP_011520959.1 | |
XM_017023659.1 | 282 | Silent Mutation | GCG,GCT | A,A 7 | XP_016879148.1 | |
XM_017023660.1 | 282 | Silent Mutation | GCG,GCT | A,A 7 | XP_016879149.1 | |
XM_017023661.1 | 282 | Silent Mutation | GCG,GCT | A,A 7 | XP_016879150.1 | |
XM_017023662.1 | 282 | UTR 5 | XP_016879151.1 |