Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGTAGGATTGATGCAAGTCACTTC[C/T]AGGAACTCCCCAACAAGCTTAATGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604230 MIM: 601421 | ||||||||||||||||||||
Literature Links: |
ADAT1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ADAT1 - adenosine deaminase, tRNA specific 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KARS - lysyl-tRNA synthetase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130089.1 | 1496 | Silent Mutation | CTA,CTG | L,L 480 | NP_001123561.1 | |
NM_005548.2 | 1496 | Intron | NP_005539.1 | |||
XM_017023217.1 | 1496 | Silent Mutation | CTA,CTG | L,L 296 | XP_016878706.1 |