Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTGATGACGGTGCCTGGCACCTCG[C/T]TCCCCACACGGCCCACTCTCCCTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607618 MIM: 609707 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DDX28 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DDX28 - DEAD-box helicase 28 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018380.3 | 2143 | Missense Mutation | NP_060850.2 |
DUS2 - dihydrouridine synthase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC100131303 - uncharacterized LOC100131303 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |