Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTCAGACTCTGTCTCTGACGCAGA[C/T]GTGAGAGGTCGGAGTGGCAGGAACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609925 MIM: 609926 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DPEP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DPEP2 - dipeptidase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001324159.1 | 622 | Missense Mutation | CAT,CGT | H,R 274 | NP_001311088.1 | |
NM_022355.3 | 622 | Missense Mutation | CAT,CGT | H,R 432 | NP_071750.1 | |
XM_005256090.4 | 622 | UTR 3 | XP_005256147.2 | |||
XM_011523266.1 | 622 | Missense Mutation | CAT,CGT | H,R 445 | XP_011521568.1 | |
XM_011523268.2 | 622 | Missense Mutation | CAT,CGT | H,R 437 | XP_011521570.1 | |
XM_011523271.2 | 622 | UTR 3 | XP_011521573.1 | |||
XM_011523272.2 | 622 | Missense Mutation | CAT,CGT | H,R 287 | XP_011521574.1 | |
XM_011523273.2 | 622 | Missense Mutation | CAT,CGT | H,R 287 | XP_011521575.1 | |
XM_011523274.2 | 622 | Missense Mutation | CAT,CGT | H,R 237 | XP_011521576.1 | |
XM_017023541.1 | 622 | Missense Mutation | CAT,CGT | H,R 379 | XP_016879030.1 | |
XM_017023542.1 | 622 | Missense Mutation | CAT,CGT | H,R 355 | XP_016879031.1 | |
XM_017023543.1 | 622 | Missense Mutation | CAT,CGT | H,R 329 | XP_016879032.1 | |
XM_017023544.1 | 622 | Missense Mutation | CAT,CGT | H,R 322 | XP_016879033.1 | |
XM_017023545.1 | 622 | Missense Mutation | CAT,CGT | H,R 287 | XP_016879034.1 | |
XM_017023546.1 | 622 | Missense Mutation | CAT,CGT | H,R 196 | XP_016879035.1 | |
XM_017023547.1 | 622 | Missense Mutation | CAT,CGT | H,R 196 | XP_016879036.1 |
DPEP3 - dipeptidase 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |