Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCTGCCCCTGGAGAGTGGAGTGTC[A/G]GACTTCAAACCTGGACAGTCTGAGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613346 | ||||||||||||||||||||
Literature Links: |
SHISA9 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SHISA9 - shisa family member 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145204.2 | 1088 | Intron | NP_001138676.2 | |||
NM_001145205.1 | 1088 | Missense Mutation | AGA,GGA | R,G 215 | NP_001138677.2 | |
XM_005255539.3 | 1088 | Intron | XP_005255596.2 | |||
XM_011522642.2 | 1088 | Intron | XP_011520944.1 |