Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCAGGTGCTCGCTGCGCCGGAAGG[C/T]CTTGCCGCAGTCGCTGCACACGAAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZSCAN10 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZSCAN10 - zinc finger and SCAN domain containing 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282415.1 | 1515 | Missense Mutation | ACC,GCC | T,A 136 | NP_001269344.1 | |
NM_001282416.1 | 1515 | Missense Mutation | ACC,GCC | T,A 393 | NP_001269345.1 | |
NM_032805.2 | 1515 | Missense Mutation | ACC,GCC | T,A 530 | NP_116194.2 | |
XM_017023791.1 | 1515 | Missense Mutation | ACC,GCC | T,A 530 | XP_016879280.1 | |
XM_017023792.1 | 1515 | Missense Mutation | ACC,GCC | T,A 498 | XP_016879281.1 |