Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCGCCAGTGCCTGCTCTGGTTTT[C/G]TGGAATGAGCAGAGGTGGGGTGGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 182381 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C16orf58 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C16orf58 - chromosome 16 open reading frame 58 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022744.3 | 1759 | UTR 3 | NP_073581.2 | |||
XM_017023570.1 | 1759 | UTR 3 | XP_016879059.1 |
LOC105371171 - uncharacterized LOC105371171 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC5A2 - solute carrier family 5 member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003041.3 | 1759 | Missense Mutation | TCT,TGT | S,C 615 | NP_003032.1 | |
XM_006721072.3 | 1759 | Missense Mutation | TCT,TGT | S,C 648 | XP_006721135.2 |