Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGCCTGGAAGGTCAGGGAAACATC[A/C]GGGAGCTGCTCTCCCAGGGGCAGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605220 MIM: 608273 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
APOBR PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
APOBR - apolipoprotein B receptor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
IL27 - interleukin 27 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145659.3 | 287 | Silent Mutation | CCG,CCT | P,P 88 | NP_663634.2 | |
XM_011545780.2 | 287 | Silent Mutation | CCG,CCT | P,P 90 | XP_011544082.1 |