Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCACCCTCTGGCCCCAGAATCTCC[C/T]TGGTCTCGTTCCAGGCCCCCCAGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603870 MIM: 612222 MIM: 610970 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CBFA2T3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CBFA2T3 - CBFA2/RUNX1 translocation partner 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GALNS - galactosamine (N-acetyl)-6-sulfatase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PABPN1L - poly(A) binding protein nuclear 1 like (cytoplasmic) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080487.2 | 164 | Missense Mutation | AAG,AGG | K,R 37 | NP_001073956.2 | |
NM_001294328.1 | 164 | Missense Mutation | AAG,AGG | K,R 37 | NP_001281257.1 | |
XM_017023230.1 | 164 | Missense Mutation | AAG,AGG | K,R 37 | XP_016878719.1 |
TRAPPC2L - trafficking protein particle complex 2-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |