Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCAGCCACACATCCAGATTAAAA[A/G]CCAGGAAGCACAGCAAACGTCGAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613588 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLEC3A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CLEC3A - C-type lectin domain family 3 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001244755.1 | 166 | Intron | NP_001231684.1 | |||
NM_005752.4 | 166 | Missense Mutation | ACC,GCC | T,A 38 | NP_005743.4 |