Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACTCCCCACATTTTTCAAGAATTT[A/C]TCTATAATCCCCATGTTCTAAGCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612969 MIM: 604191 MIM: 601473 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TIGD7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TIGD7 - tigger transposable element derived 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033208.3 | 2887 | Nonsense Mutation | GAA,TAA | E,* 422 | NP_149985.2 |
ZNF263 - zinc finger protein 263 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005741.4 | 2887 | Intron | NP_005732.2 | |||
XM_006720831.1 | 2887 | Intron | XP_006720894.1 | |||
XM_006720832.1 | 2887 | Intron | XP_006720895.1 | |||
XM_006720833.1 | 2887 | Intron | XP_006720896.1 | |||
XM_006720834.3 | 2887 | Intron | XP_006720897.1 | |||
XM_011522344.1 | 2887 | Intron | XP_011520646.1 | |||
XM_017022850.1 | 2887 | Intron | XP_016878339.1 |
ZNF75A - zinc finger protein 75a | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |