Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGAGGACAGAGCCCCGGCCGCAG[C/G]GGGCAGCTCACTGGTCAGGGTGTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604567 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DOC2A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DOC2A - double C2 domain alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282062.1 | 1366 | Missense Mutation | CCT,GCT | P,A 392 | NP_001268991.1 | |
NM_001282063.1 | 1366 | Missense Mutation | CCT,GCT | P,A 392 | NP_001268992.1 | |
NM_001282068.1 | 1366 | Missense Mutation | CCT,GCT | P,A 392 | NP_001268997.1 | |
NM_003586.2 | 1366 | Missense Mutation | CCT,GCT | P,A 392 | NP_003577.2 | |
XM_011545975.1 | 1366 | Missense Mutation | CCT,GCT | P,A 392 | XP_011544277.1 | |
XM_017023776.1 | 1366 | Missense Mutation | CCT,GCT | P,A 392 | XP_016879265.1 | |
XM_017023777.1 | 1366 | Missense Mutation | CCT,GCT | P,A 392 | XP_016879266.1 | |
XM_017023778.1 | 1366 | Missense Mutation | CCT,GCT | P,A 274 | XP_016879267.1 |
INO80E - INO80 complex subunit E | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304562.1 | 1366 | Intron | NP_001291491.1 | |||
NM_001304563.1 | 1366 | Intron | NP_001291492.1 | |||
NM_173618.2 | 1366 | Intron | NP_775889.1 | |||
XM_011545809.2 | 1366 | Intron | XP_011544111.1 | |||
XM_011545811.2 | 1366 | Intron | XP_011544113.1 | |||
XM_011545812.2 | 1366 | Intron | XP_011544114.1 | |||
XM_017023169.1 | 1366 | Intron | XP_016878658.1 |