Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGAAATTCCACACAGGTACCACA[A/G]GATATTATGCATGCTCCAATCGAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605943 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GDE1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GDE1 - glycerophosphodiester phosphodiesterase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001324066.1 | 957 | Silent Mutation | CTG,TTG | L,L 155 | NP_001310995.1 | |
NM_001324067.1 | 957 | Silent Mutation | CTG,TTG | L,L 234 | NP_001310996.1 | |
NM_016641.3 | 957 | Silent Mutation | CTG,TTG | L,L 265 | NP_057725.1 |
TMC5 - transmembrane channel like 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |